COM448613364 Combi-Blocks, Inc.

{4-[5-(Thiophen-2-yl)-1,2,4-oxadiazol-3-yl]phenyl}boronic acid

  • MDL number MFCD27935842
  • Cas Number : 2377608-39-2
  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0677

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0677
Availability details for
Quantity
  • Availability
  • Add to Cart