COM448613365 Combi-Blocks, Inc.

{1-[(4-Methoxyphenyl)methyl]-1,3-benzodiazol-5-yl}boronic acid

  • MDL number MFCD27935843
  • Cas Number : 2377609-78-2
  • Molecular Weight : 282.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0678

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0678
Availability details for
Quantity
  • Availability
  • Add to Cart