COM448613366 Combi-Blocks, Inc.

{3-[(4S)-4-tert-Butyl-4,5-dihydro-1,3-oxazol-2-yl]phenyl}boronic acid

  • MDL number MFCD27935844
  • Molecular Weight : 247.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0679

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0679
Availability details for
Quantity
  • Availability
  • Add to Cart