COM448613374 Combi-Blocks, Inc.

3-(1,1-Dioxo-1,2-thiazolidin-2-yl)-5-(trifluoromethyl)phenylboronic acid

  • MDL number MFCD27935852
  • Molecular Weight : 309.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0687

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0687
Availability details for
Quantity
  • Availability
  • Add to Cart