COM448613379 Combi-Blocks, Inc.

[4-(4-Butylpiperazine-1-sulfonyl)phenyl]boronic acid

  • MDL number MFCD27935857
  • Molecular Weight : 326.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0692
Availability details for
Quantity
  • Availability
  • Add to Cart