COM448613383 Combi-Blocks, Inc.

[5-(5-Phenyl-1,3,4-oxadiazol-2-yl)pyridin-3-yl]boronic acid

  • MDL number MFCD27935860
  • Molecular Weight : 267.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0696

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0696
Availability details for
Quantity
  • Availability
  • Add to Cart