COM448613386 Combi-Blocks, Inc.

[3-(Dimethylsulfamoyl)-5-(trifluoromethyl)phenyl]boronic acid

  • MDL number MFCD27935863
  • Molecular Weight : 297.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0699

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0699
Availability details for
Quantity
  • Availability
  • Add to Cart