COM448613390 Combi-Blocks, Inc.

{4-[5-(4-Methylphenyl)-1,2,4-oxadiazol-3-yl]phenyl}boronic acid

  • MDL number MFCD27935867
  • Cas Number : 2377609-92-0
  • Molecular Weight : 280.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0703

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0703
Availability details for
Quantity
  • Availability
  • Add to Cart