COM448613393 Combi-Blocks, Inc.

{4-[4-(2-Methylpropyl)piperazine-1-sulfonyl]phenyl}boronic acid

  • MDL number MFCD27935870
  • Molecular Weight : 326.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0706

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0706
Availability details for
Quantity
  • Availability
  • Add to Cart