COM448613394 Combi-Blocks, Inc.

{1-[4-(Dimethylsulfamoyl)phenyl]pyrazol-4-yl}boronic acid

  • MDL number MFCD27935871
  • Molecular Weight : 295.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0707

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0707
Availability details for
Quantity
  • Availability
  • Add to Cart