COM448613407 Combi-Blocks, Inc.

{1-[4-(Diethylsulfamoyl)phenyl]pyrazol-4-yl}boronic acid

  • MDL number MFCD27935884
  • Molecular Weight : 323.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0720

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0720
Availability details for
Quantity
  • Availability
  • Add to Cart