COM448613411 Combi-Blocks, Inc.

{1-[4-(Pyrrolidine-1-sulfonyl)phenyl]pyrazol-4-yl}boronic acid

  • MDL number MFCD27935888
  • Molecular Weight : 321.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0724

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0724
Availability details for
Quantity
  • Availability
  • Add to Cart