COM448613416 Combi-Blocks, Inc.

[3,5-Bis(benzyloxy)phenyl]boronic acid

  • MDL number MFCD27935893
  • Cas Number : 906665-99-4
  • Molecular Weight : 334.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0729

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0729
Availability details for
Quantity
  • Availability
  • Add to Cart