COM448613419 Combi-Blocks, Inc.

[3-(Adamantan-1-yl)-4-(benzyloxy)phenyl]boronic acid

  • MDL number MFCD27935896
  • Cas Number : 221137-01-5
  • Molecular Weight : 362.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0734

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0734
Availability details for
Quantity
  • Availability
  • Add to Cart