COM448613422 Combi-Blocks, Inc.

6-Carboxy-2,3-difluorophenylboronic acid

  • MDL number MFCD28127177
  • Cas Number : 2377609-91-9
  • Molecular Weight : 201.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-1505

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-1505
Availability details for
Quantity
  • Availability
  • Add to Cart