COM448613424 Combi-Blocks, Inc.

4-(4-Oxopiperidine-1-sulfonyl)phenylboronic acid

  • MDL number MFCD28024837
  • Molecular Weight : 283.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-1545

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-1545
Availability details for
Quantity
  • Availability
  • Add to Cart