COM448613425 Combi-Blocks, Inc.

4-Chloro-3-(hydroxymethyl)phenylboronic acid

  • MDL number MFCD28122667
  • Cas Number : 1430237-54-9
  • Molecular Weight : 186.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-1573

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-1573
Availability details for
Quantity
  • Availability
  • Add to Cart