COM448613429 Combi-Blocks, Inc.

3-Amino-5-nitrophenylboronic acid, pinacol ester

  • MDL number MFCD07363740
  • Cas Number : 871329-51-0
  • Molecular Weight : 264.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2004

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2004
Availability details for
Quantity
  • Availability
  • Add to Cart