COM448613437 Combi-Blocks, Inc.

3-(N-Boc-aminomethyl)phenylboronic acid

  • MDL number MFCD06246052
  • Cas Number : 199609-62-6
  • Molecular Weight : 251.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2013

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2013
Availability details for
Quantity
  • Availability
  • Add to Cart