COM448613439 Combi-Blocks, Inc.

(3-Methanesulfonylaminomethyl)phenylboronic acid

  • MDL number MFCD06659817
  • Cas Number : 850568-39-7
  • Molecular Weight : 229.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2016

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2016
Availability details for
Quantity
  • Availability
  • Add to Cart