COM448613440 Combi-Blocks, Inc.

4-Isopropoxy-1,3-phenylenediboronic acid

  • MDL number MFCD04115638
  • Cas Number : 850568-40-0
  • Molecular Weight : 223.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2017

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2017
Availability details for
Quantity
  • Availability
  • Add to Cart