COM448613442 Combi-Blocks, Inc.

3-Acetamidomethylphenylboronic acid

  • MDL number MFCD06659819
  • Cas Number : 850568-42-2
  • Molecular Weight : 193.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2019

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2019
Availability details for
Quantity
  • Availability
  • Add to Cart