COM448613450 Combi-Blocks, Inc.

4-TBDMS-hydroxymethylphenylboronic acid

  • MDL number MFCD07368897
  • Cas Number : 162356-89-0
  • Molecular Weight : 266.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2030

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2030
Availability details for
Quantity
  • Availability
  • Add to Cart