COM448613470 Combi-Blocks, Inc.

4-(2-Carboxyethyl)phenylboronic acid

  • MDL number MFCD01318119
  • Cas Number : 166316-48-9
  • Molecular Weight : 194.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2055

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2055
Availability details for
Quantity
  • Availability
  • Add to Cart