COM448613473 Combi-Blocks, Inc.

3-(2-Methoxycarbonylethyl)phenylboronic acid

  • MDL number MFCD04115648
  • Cas Number : 833472-82-5
  • Molecular Weight : 208.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2066

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2066
Availability details for
Quantity
  • Availability
  • Add to Cart