COM448613474 Combi-Blocks, Inc.

3-(2-Carboxyethyl)phenylboronic acid

  • MDL number MFCD04115649
  • Cas Number : 693803-17-7
  • Molecular Weight : 194.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2067

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2067
Availability details for
Quantity
  • Availability
  • Add to Cart