COM448613477 Combi-Blocks, Inc.

4-(3-Hydroxypropyl)phenylboronic acid

  • MDL number MFCD04973734
  • Cas Number : 850568-48-8
  • Molecular Weight : 180.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2072

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2072
Availability details for
Quantity
  • Availability
  • Add to Cart