COM448613481 Combi-Blocks, Inc.

4-(2-Ethoxycarbonylethyl)phenylboronic acid

  • MDL number MFCD04115652
  • Cas Number : 660440-57-3
  • Molecular Weight : 222.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2077

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2077
Availability details for
Quantity
  • Availability
  • Add to Cart