COM448613538 Combi-Blocks, Inc.

2-(tert-Butylcarbamoyl)-5-methoxyphenylboronic acid

  • MDL number MFCD22375009
  • Cas Number : 2096333-56-9
  • Molecular Weight : 251.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2146

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2146
Availability details for
Quantity
  • Availability
  • Add to Cart