COM448613541 Combi-Blocks, Inc.

(4-Benzyloxycarbonyl-2-nitro)phenylboronic acid

  • MDL number MFCD02179461
  • Cas Number : 850568-58-0
  • Molecular Weight : 301.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2149

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2149
Availability details for
Quantity
  • Availability
  • Add to Cart