COM448613552 Combi-Blocks, Inc.

3-Amino-5-carboxylphenylboronic acid

  • MDL number MFCD02179466
  • Cas Number : 116378-40-6
  • Molecular Weight : 181.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2160

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2160
Availability details for
Quantity
  • Availability
  • Add to Cart