COM448613555 Combi-Blocks, Inc.

2-Amino-4-carboxylphenylboronic acid , HCl

  • MDL number MFCD06656012
  • Cas Number : 850568-60-4
  • Molecular Weight : 217.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2163

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2163
Availability details for
Quantity
  • Availability
  • Add to Cart