COM448613559 Combi-Blocks, Inc.

2-(E-3-Methoxy-3-oxo-1-propen-1-yl)phenylboronic acid

  • MDL number MFCD02179475
  • Cas Number : 372193-68-5
  • Molecular Weight : 206.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2167

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2167
Availability details for
Quantity
  • Availability
  • Add to Cart