COM448613560 Combi-Blocks, Inc.

4-(E-3-Methoxy-3-oxo-1-propen-1-yl)phenylboronic acid

  • MDL number MFCD02179476
  • Cas Number : 380430-58-0
  • Molecular Weight : 206.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2168

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2168
Availability details for
Quantity
  • Availability
  • Add to Cart