COM448613561 Combi-Blocks, Inc.

3-(E-3-Methoxy-3-oxo-1-propen-1-yl)phenylboronic acid

  • MDL number MFCD02179477
  • Cas Number : 380430-59-1
  • Molecular Weight : 206.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2169

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2169
Availability details for
Quantity
  • Availability
  • Add to Cart