COM448613570 Combi-Blocks, Inc.

2,2'-Bithiophene-5-boronic acid

  • MDL number MFCD03086112
  • Cas Number : 132898-95-4
  • Molecular Weight : 210.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2181

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2181
Availability details for
Quantity
  • Availability
  • Add to Cart