COM448613571 Combi-Blocks, Inc.

4-Chloro-2-ethoxycarbonylphenylboronic acid

  • MDL number MFCD06659823
  • Cas Number : 850568-61-5
  • Molecular Weight : 228.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2183

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2183
Availability details for
Quantity
  • Availability
  • Add to Cart