COM448613584 Combi-Blocks, Inc.

2-Chloro-4-trifluoromethoxyphenylboronic acid

  • MDL number MFCD11040272
  • Cas Number : 345226-20-2
  • Molecular Weight : 240.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2198

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2198
Availability details for
Quantity
  • Availability
  • Add to Cart