COM448613598 Combi-Blocks, Inc.

3-Isocyanatophenylboronic acid, pinacol ester

  • MDL number MFCD06659827
  • Cas Number : 787591-43-9
  • Molecular Weight : 245.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2213

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2213
Availability details for
Quantity
  • Availability
  • Add to Cart