COM448613603 Combi-Blocks, Inc.

3-Acetamido-4-nitrophenylboronic acid

  • MDL number MFCD12025959
  • Cas Number : 78887-37-3
  • Molecular Weight : 224.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2218

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2218
Availability details for
Quantity
  • Availability
  • Add to Cart