COM448613666 Combi-Blocks, Inc.

4-Benzyloxy-3-formylphenylboronic acid

  • MDL number MFCD02179487
  • Cas Number : 121124-98-9
  • Molecular Weight : 256.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2307

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2307
Availability details for
Quantity
  • Availability
  • Add to Cart