COM448613699 Combi-Blocks, Inc.

3-(Piperidin-1-ylsulfonyl)phenylboronic acid

  • MDL number MFCD07783850
  • Cas Number : 690662-96-5
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2349

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2349
Availability details for
Quantity
  • Availability
  • Add to Cart