COM448613729 Combi-Blocks, Inc.

5-Methyl-4-propoxy-1,3-phenylenediboronic acid

  • MDL number MFCD06659835
  • Cas Number : 850568-70-6
  • Molecular Weight : 237.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2389

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2389
Availability details for
Quantity
  • Availability
  • Add to Cart