COM448613733 Combi-Blocks, Inc.

5-Methylpyridine-2-boronic acid

  • MDL number MFCD00674179
  • Cas Number : 372963-49-0
  • Molecular Weight : 136.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2393

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2393
Availability details for
Quantity
  • Availability
  • Add to Cart