COM448613743 Combi-Blocks, Inc.

2-(Hydroxymethyl)-6-methoxyphenylboronic acid dehydrate

  • MDL number MFCD16036102
  • Cas Number : 947163-27-1
  • Molecular Weight : 164.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2407

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2407
Availability details for
Quantity
  • Availability
  • Add to Cart