COM448613751 Combi-Blocks, Inc.

N,N-Diethyl 4-boronobenzenesulfonamide

  • MDL number MFCD06659846
  • Cas Number : 850568-76-2
  • Molecular Weight : 257.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2420

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2420
Availability details for
Quantity
  • Availability
  • Add to Cart