COM448613753 Combi-Blocks, Inc.

4-(Cyclopentyloxy)-2,3-difluorophenylboronic acid

  • MDL number MFCD19981508
  • Cas Number : 2096331-20-1
  • Molecular Weight : 242.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2422

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2422
Availability details for
Quantity
  • Availability
  • Add to Cart