COM448613759 Combi-Blocks, Inc.

N-Butyl 4-boronobenzenesulfonamide

  • MDL number MFCD07363753
  • Cas Number : 850589-32-1
  • Molecular Weight : 257.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2428

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2428
Availability details for
Quantity
  • Availability
  • Add to Cart