COM448613763 Combi-Blocks, Inc.

4-(Thiomorpholinylsulfonyl)phenylboronic acid

  • MDL number MFCD07363757
  • Cas Number : 871329-69-0
  • Molecular Weight : 287.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2432

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2432
Availability details for
Quantity
  • Availability
  • Add to Cart