COM448613797 Combi-Blocks, Inc.

2-Methyl-3-nitrophenylboronic acid

  • MDL number MFCD06201025
  • Cas Number : 1072945-60-8
  • Molecular Weight : 181.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2474

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2474
Availability details for
Quantity
  • Availability
  • Add to Cart